Orders LinkContactUsLinkComponent
Precolumn filters
Precolumn filter, Rustfrit stål, 2,0 µm, 0,68 µl, 1000 bar
  JOURJR-69230-2
Precolumn filters
  JOURJR-69230-2
Precolumn filters
Precolumn filter, Rustfrit stål, 2,0 µm, 0,68 µl, 1000 bar
2.200,00 kr
1 * 1 Items
  • Porøsitet:
    2,0 µm
  • Beskrivelse:
    Precolumn filter
  • Materiale:
    Rustfrit stål
  • Seq:
    08
  • Slagvolumen:
    0,68 µl
  • Tryk:
    1000 bar