Cut resistant gloves, Sharpflex PU
8, grøn/sort, 250 mm
Katalog # DALL2232523/8
Leverandør: HONEYWELL SAFETY
Gå til:
Product Details & Documents
These gloves knitted from composite and para-aramid yarn features a black PU coating on the palm.
- Knitting technique provides the glove with excellent fit and high dexterity
- Para-aramid yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fiber
- PU coating increases the abrasion resistance and allows use in slightly greasy environments
Ideal for handling heavy parts with high cut risks in dry and slightly greasy environments.
Certifications: EN 388:2016 (4X41C); Cat. II
Specifikationer
- Handske størrelse:8
- Farve:grøn/sort
- Længde:250 mm
- Seq:02
- Cat. No.:DALL2232523/8
- Foringsmateriale:Para-aramid
- Ambidextrous/Hånd-specifik:Hånd-specifik
- Model:PU
- Manchet:Strikket
- EN norm:EN 388:2016 (4X41C)
- Overflade:Coating on palm and fingertips
- Engangs/Genbrugelig:Genanvendelig
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



