White light viewer, LabLite illuminator, ViewOne™
Product Details & Documents
Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
- Uniform background for enhanced visualization of colorimetric samples
- Waterproof top allows for visualizing wet samples such as SDS-PAGE gels or dissection samples
- Portable and space saving
The ViewOne™ LabLite is a stand-alone bench top solution for visualizing colorimetric sample against a uniform white light background. Labs have used the ViewOne with ELISA plates, colony picking and bacterial plates, hemagglutiation assays and cell culture plates. The waterproof top provides a protected surface for usage with set samples such as Coomassie stained SDS-PAGE gels, zymogram gels, and samples for dissection.
The ViewOne™ LabLite Set includes a tilt stand that offers relief from loading over a plate, and adjusts to 10°, 30°, 45°, making it a practical unit that can also be used in a biosafety cabinet.
Leveringsinformation: The ViewOne™ LabLite set includes VL-1007 Universal tilt stand for viewing angle adjustment. Stand-alone model does not include tilt stand.
Specifikationer
- Model:VL-1200
- Beskrivelse:ViewOne™ LabLite illuminator set, for 96-well ELISA plate, Petri dish, SDS-PAGE gel, 384-well plates
- Dimensioner:115×170×5 mm
- Seq:02