Orders LinkContactUsLinkComponent
Adapters, delivery, straight
Adapters, delivery, straight
29/32 NS, 100 mm
Katalog # WITG1.900.029
Leverandør:   witeg Labortechnik
Adapters, delivery, straight
Katalog # WITG1.900.029
Leverandør:  witeg Labortechnik
Adapters, delivery, straight
Adapters, delivery, straight
29/32 NS, 100 mm
Price
263,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifikationer
  • Kappestr.:
    29/32 NS
  • Rørlængde:
    100 mm