Cut resistant gloves, Sharpflex Nit
10, grøn/sort, 250 mm
DALL2232524/10
These gloves knitted from composite and para-aramid yarn features a black nitrile foam coating on the palm.
- Knitting technique provides the glove with excellent fit and high dexterity
- Para-aramid yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fiber
- Nitrile coating provides good protection against oil and grease
- Foam coating is more flexible than standard nitrile coating
Ideal for handling heavy parts with high cut risks in oily and greasy environments.
: EN 388:2016 (3X42C), EN 407 (X1XXXX); Cat. II
- Handske størrelse:10
- Farve:grøn/sort
- Længde:250 mm
- Seq:04
- Cat. No.:DALL2232524/10
- Foringsmateriale:Para-aramid
- Ambidextrous/Hånd-specifik:Hånd-specifik
- Måler:13
- Model:Nit
- Manchet:Strikket
- EN norm:EN 388:2016 (3X42C), EN 407 (X1XXXX)
- Overflade:Foam coating on palm and fingertips
- Engangs/Genbrugelig:Genanvendelig