Phenolphthalein ACS
Glass bottle for solids, 50 g
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS nummer:77-09-8
- Formel:C₂₀H₁₄O₄
- MDL nummer:MFCD00005913
- Molecular Weight:318.33 g/mol
- Merck Index:14,07243
- Smeltende Pt:258…262 °C
- Tæthed:1,3 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-004-7
- NÅ:01-2119498295-24
Documents
Specifikationer
Specifikationstestresultater
- Clarity of Alcohol Solution:Pass
- Visual Transition Interval (pH 8 – 10):Pass
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



