Phenolphthalein ACS
Glass bottle for solids, 100 g
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS nummer:77-09-8
- Formel:C₂₀H₁₄O₄
- MDL nummer:MFCD00005913
- Molecular Weight:318.33 g/mol
- Merck Index:14,07243
- Smeltende Pt:258…262 °C
- Tæthed:1,3 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-004-7
- NÅ:01-2119498295-24
Specifikationer
Specifikationstestresultater
- Clarity of Alcohol Solution:Pass
- Visual Transition Interval (pH 8 – 10):Pass
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



