Gentamicin sulfat
Glass bottle, 5 g
Katalog # TCIAG0383-5G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
- CAS nummer:1405-41-0
- MDL nummer:MFCD00270181
- Merck Index:13,04403
- Smeltende Pt:218…237 °C
- Opbevaringstemperatur:Køleskab
- EINECS:215-778-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Light yellow powder to crystal
- Total Nitrogen:8.0 to 10.0 %
- Specific rotation [a]20/D:+107.0 to +121.0 deg (C=1, H2O) (calcd.on anh.substance)
- Water:max. 18.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



