Accessories
Silicone protection cover, MSAH352, shields the microscope from dirt and stains, compatible with all Dino-Lite models
Katalog # IDCPMSAH352
Leverandør: Dino-Lite / IDCP B.V.
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Silicone protection cover, MSAH352, shields the microscope from dirt and stains, compatible with all Dino-Lite models
- Cat. No.:IDCPMSAH352