Handheld digital microscopes, Dino-Lite Special Light, UV
UV microscope, with emission filter, AM4113FVT, 10 - 70×, 200×, 8 LEDs (390/400 nm UV), 1,3 MP
IDCPAM4113FVT
These handheld microscopes feature ultraviolet (UV) light or a combination between UV and white light.
- High speed USB 2.0 output
- Metal housing or composite housing; metal models are ESD-safe
- Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
- Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
- Integrated polariser greatly reduces glare on reflective surfaces (some models only)
- Exchangeable caps for more flexibility (some models only)
: Supplied with carry pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), user manual, Quick guide and calibration target.
- Forstørrelse:10 - 70×, 200×
- Model:AM4113FVT
- Opløsning:1,3 MP
- Beskrivelse:UV microscope, with emission filter
- Seq:05
- Belysning:8 LEDs (390/400 nm UV)
Accessories
- BeskrivelseB×D×H
- IDCPRK-06ATabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX150×220×270 mm1 * 1 Items - 1.880,00 kr
- Beskrivelse
- 630-2923Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery1 * 1 Items - 2.460,00 kr
- Beskrivelse
- Beskrivelse
- 630-2517Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm1 * 1 Items - 2.540,00 kr