Grinding Jars
50 ml, Rustfrit stål, CryoMill
Katalog # 412-0226
Leverandør: Retsch GmbH
Gå til:
Product Details & Documents
These grinding jars help to provide efficient milling.
Specifikationer
- Til:CryoMill
- Kapacitet:50 ml
- Materiale:Rustfrit stål
- Seq:12