Grinding Jars
25 ml, Zirconiumoxid, CryoMill
Katalog # 412-0230
Leverandør: Retsch GmbH
Gå til:
Product Details & Documents
These grinding jars help to provide efficient milling.
Specifikationer
- Til:CryoMill
- Kapacitet:25 ml
- Materiale:Zirconiumoxid