Orders LinkContactUsLinkComponent
Measuring scoops
15 ml, 115 mm, ufarvet
Catalog # HIRS7780101
Supplier:   HIRSCHMANN
Measuring scoops
Catalog # HIRS7780101
Supplier:  HIRSCHMANN
Measuring scoops
15 ml, 115 mm, ufarvet
Price
119,00 kr
Log in for availability
1 * 12 Items
Product Details & Documents
HDPE, natural colour.
Specifications
  • Kapacitet:
    15 ml
  • Farve:
    ufarvet
  • Længde:
    115 mm
  • Seq:
    01