Pipette tips for labopette®
Multibox, autoclavable, refill trays of pipette tips up to 300 µl
Katalog # HIRS9490109
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifikationer
- Seq:16
- Spidstype:Multibox, autoclavable
- Til brug med:refill trays of pipette tips up to 300 µl