Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test Strips, Oil

Test Strips, Oil

QUANTOFIX® nitrite/pH, 0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 6,0 • 6,4 • 6,7 • 7,0 • 7,3 • 7,6 • 7,9 • 8,2 • 8,4 • 8,6 • 8,8 • 9,0 • 9,3 • 9,6, Nitrite: Azo reaction with an aromatic amine; pH: pH indicator, 43×123×40 mm (packaging), 100 Tests, 100 Tests
Katalog # MANA91338
Leverandør:   MACHEREY-NAGEL
Test Strips, Oil
Katalog # MANA91338
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
Test Strips, Oil
Test Strips, Oil

QUANTOFIX® nitrite/pH, 0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 6,0 • 6,4 • 6,7 • 7,0 • 7,3 • 7,6 • 7,9 • 8,2 • 8,4 • 8,6 • 8,8 • 9,0 • 9,3 • 9,6, Nitrite: Azo reaction with an aromatic amine; pH: pH indicator, 43×123×40 mm (packaging), 100 Tests, 100 Tests

CAS nummer:  
Price
417,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

This test paper allows the rapid and reliable detection of oil contamination of water and soil.

  • Colour changes: Blue to dark blue

For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample.

Specifikationer

Specifikationer

  • Kemisk metode:
    Nitrite: Azo reaction with an aromatic amine; pH: pH indicator
  • Pakn:
    100 Tests
  • Beskrivelse:
    QUANTOFIX® nitrite/pH
  • Dimensioner:
    43×123×40 mm (packaging)
  • Test pr. sæt:
    100 Tests
  • Område:
    0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 6,0 • 6,4 • 6,7 • 7,0 • 7,3 • 7,6 • 7,9 • 8,2 • 8,4 • 8,6 • 8,8 • 9,0 • 9,3 • 9,6
  • Indhold:
    100 test strips in a tube, instruction leaflet
  • Cat. No.:
    MANA91338
  • Overensstemmelse:
    No hazardous material
  • Platform:
    Semi-quantitative test strips
  • Holdbarhed (måneder):
    30 (from production)
  • Vægt:
    35 g
  • Instrument:
    No
  • Parameter:
    All reagents included
  • Method:
    Nitrite - Griess reaction (white → red-violet);pH - mixed indicator (yellow-orange → violet-red)

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.