Micro extraction by packed sorbent (MEPS™)
XCHANGE® syringe, 250 µl, MEPS™ BINs
Katalog # SGEA2928600
Leverandør: Trajan Scientific and Medical
Micro extraction by packed sorbent (MEPS™)
Katalog # SGEA2928600
Leverandør: Trajan Scientific and Medical
Gå til:
Product Details & Documents
Micro Extraction by Packed Sorbent (MEPS™) is a new development in the field of sample preparation by SPE (solid phase extraction). The MEPS™ product consists of two parts, the MEPS™ syringe and the MEPS™ Barrel Insert and Needle assembly (BIN). The MEPS™ approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries. Because MEPS™ BINs can be used for sample volumes as small as 10 µl it makes them particularly well suited to on-line use with LC-MS analysis of volume limited samples.
- The BIN contains the SPE phase that is built into the syringe needle
- The BIN is used with a 100 or 250 µl gas tight MEPS™ syringe that allows fluid handling at normal SPE pressures
- Sample preparation for complex biological samples is readily adapted to MEPS™ and reduces the volume of sample and reagents required for extraction
Base material is silica with mean particle size of 45 µm and pore size of 60 Å.
Specifikationer
- Til:MEPS™ BINs
- Volumen:250 µl
- Beskrivelse:XCHANGE® syringe
- Seq:07