Syringes for CTC analytics systems
Fixed needle, 26, 10 µl, 50 mm, 0,47 mm, Conical, 10F-BT-GT-5/0.47C
Katalog # SGEA0029771
Leverandør: Trajan Scientific and Medical
Gå til:
Product Details & Documents
* gastæt
Specifikationer
- Udv.Ø:0,47 mm
- Måler:26
- Model:10F-BT-GT-5/0.47C
- Spidstype:Conical
- Volumen:10 µl
- Beskrivelse:Fixed needle
- Længde:50 mm
- Seq:25