Orders LinkContactUsLinkComponent
Safety helmets, 1100
Safety helmets, 1100
Safety helmet, unvented, UK, blå
Katalog # 111-1502
Safety helmets, 1100
Katalog # 111-1502
Leverandør:  
Safety helmets, 1100
Safety helmets, 1100
Safety helmet, unvented, UK, blå
Discontinued
Product Details & Documents
Protective helmet with helmet shell made from high density polyethylene (HDPE). For use in construction and industry.
  • With six point LDPE inner cradle
  • Ear protection, face protection shield and chin strap available as accessories
Size: 510 to 630 mm
Weight: 375 g
EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)
 : EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)
Documents
Specifikationer
  • Farve:
    blå
  • Beskrivelse:
    Safety helmet, unvented, UK