Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Safety helmets, 1125

Safety helmets, 1125

Safety helmet, vented, EU, hvid
Katalog # 111-1520
Safety helmets, 1125
Katalog # 111-1520
Safety helmets, 1125
Safety helmets, 1125

Safety helmet, vented, EU, hvid

Price
101,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Impact resistant protective helmet with helmet shell made from high density polyethylene (HDPE). Suitable for the construction industry.

  • With six point terylene inner cradle
  • Front sweatband made from brushed nylon fabric
  • Ear protection, face protection shield and chin strap available as accessories

Size: 510 to 630 mm.

Weight: 365 g.

EN 397 (unvented version); Electrical Resistance and Molten Metal applicable to unvented shells only.

Certifications: EN 397 (unvented version); Electrical Resistance and Molten Metal applicable to unvented shells only.

Specifikationer
  • Farve:
    hvid
  • Beskrivelse:
    Safety helmet, vented, EU
  • Seq:
    22