Objectives for BA series
EC PL plan achromatic objective (spring), 100×/1,25
Katalog # MOTI1101001703241
Leverandør: MOTIC
Gå til:
Product Details & Documents
Specifikationer
- Forstørrelse:100×/1,25
- Beskrivelse:EC PL plan achromatic objective (spring)