Orders LinkContactUsLinkComponent
Objectives for BA series
Objectives for BA series
EC PL plan achromatic objective (spring), 100×/1,25
Katalog # MOTI1101001703241
Leverandør:   MOTIC
Objectives for BA series
Katalog # MOTI1101001703241
Leverandør:  MOTIC
Objectives for BA series
Objectives for BA series
EC PL plan achromatic objective (spring), 100×/1,25
Price
1.120,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Forstørrelse:
    100×/1,25
  • Beskrivelse:
    EC PL plan achromatic objective (spring)