Orders LinkContactUsLinkComponent
Objectives for BA series
Objectives for BA series
EC PL plan achromatic objective, 20×/0,45
Katalog # MOTI1101001703611
Leverandør:   MOTIC
Objectives for BA series
Katalog # MOTI1101001703611
Leverandør:  MOTIC
Objectives for BA series
Objectives for BA series
EC PL plan achromatic objective, 20×/0,45
Price
782,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Forstørrelse:
    20×/0,45
  • Beskrivelse:
    EC PL plan achromatic objective