Orders LinkContactUsLinkComponent
SaltLine syringes
SaltLine syringes
2500 µl, TLL-SAL calibrated
Katalog # HAMICAL203250
Leverandør:   HAMILTON BONADUZ
SaltLine syringes
Katalog # HAMICAL203250
Leverandør:  HAMILTON BONADUZ
SaltLine syringes
SaltLine syringes
2500 µl, TLL-SAL calibrated
Price
3.260,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Model:
    TLL-SAL calibrated
  • Volumen:
    2500 µl
  • Seq:
    09