Orders LinkContactUsLinkComponent
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN55PA
Katalog # MEMMUN55PAV3
Leverandør:   MEMMERT
Loop interfaces, 4 – 20 mA current
Katalog # MEMMUN55PAV3
Leverandør:  MEMMERT
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN55PA
Price
1.780,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C
  • Seq:
    074
  • Til:
    UN55PA