Orders LinkContactUsLinkComponent
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN160PA
Catalog # MEMMUN160PAV3
Supplier:   MEMMERT
Loop interfaces, 4 – 20 mA current
Catalog # MEMMUN160PAV3
Supplier:  MEMMERT
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN160PA
Price
1.780,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C
  • Seq:
    080
  • Til:
    UN160PA