Accessories
Port, Ø 14 mm, can be closed by flap, custom positions in the back wall, IN260plus
Katalog # MEMMIN260PLUSD6
Leverandør: MEMMERT
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Port, Ø 14 mm, can be closed by flap, custom positions in the back wall
- Seq:06
- Til:IN260plus