Slide mailers
Cubic slide mailers with closure, gennemsigtig, PP, 5
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Slide mailers made of polyethylene for archiving and staining of microscope slides.
- Secure closure and space for ²/₅ slides
- Break resistant rails for easy accommodation
- Transport of slides without contacting each other
Documents
Specifikationer
- Antal objektglas:5
- Farve:gennemsigtig
- Beskrivelse:Cubic slide mailers with closure
- Materiale:PP
- Seq:02