Sample containers, straight, with screw cap, WHEATON®
150 ml, Plain label, sterile, klar, PS/metal
Leverandør: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Katalog # 216-1017
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Documents
Specifikationer
- Model:Plain label, sterile
- Kapacitet:150 ml
- Farve:klar
- Materiale:PS/metal