Rectangular cover glasses, Corning®
24×60 mm, #1,5
Supplier: Corning
Rectangular cover glasses, Corning®
Catalog # CORN2980-246
Supplier: Corning
Jump to:
Product Details & Documents
These rectangular cover glasses are made from borosilicate glass.
Documents
Certificates
Search by Lot NumberSpecifications
- Antal:#1,5
- L×B:24×60 mm
- Seq:07
- Cat. No.:CORN2980-246
- Miljøcertificering:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Lav fremstillingspåvirkning