Cover glasses, Corning®
22×30 mm, 0,12 - 0,16 mm, Borosilikatglas
Supplier: Corning
Jump to:
Product Details & Documents
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.
- Designed for use in general laboratory microscopy
Nonsterile.
Documents
Certificates
Search by Lot NumberSpecifications
- L×B:22×30 mm
- Materiale:Borosilikatglas
- Tykkelse:0,12 - 0,16 mm
- Cat. No.:CORN2975-223
- Miljøcertificering:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Lav fremstillingspåvirkning