Cover glasses, Corning®
24×50 mm, 0,12 - 0,16 mm, Borosilikatglas
: Corning
- L×B:24×50 mm
- Materiale:Borosilikatglas
- Tykkelse:0,12 - 0,16 mm
- Cat. No.:CORN2975-245
- Miljøcertificering:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Lav fremstillingspåvirkning