Orders LinkContactUsLinkComponent
Cover glasses, Corning®
Cover glasses, Corning®
24×60 mm, 0,12 - 0,16 mm, Borosilikatglas
  CORN2975-246
 :   Corning
Cover glasses, Corning®
  CORN2975-246
 :  Corning
Cover glasses, Corning®
Cover glasses, Corning®
24×60 mm, 0,12 - 0,16 mm, Borosilikatglas
3.420,00 kr
1 * 1.000 Items
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.
  • Designed for use in general laboratory microscopy
Nonsterile.
Documents
  • L×B:
    24×60 mm
  • Materiale:
    Borosilikatglas
  • Tykkelse:
    0,12 - 0,16 mm