Silikoneslanger
Size 16, with hose barb of 3,1 mm, 3,00 mm, 16
Catalog # SPECACTU-P16-25N
Supplier: Spectrum Laboratories
Jump to:
Product Details & Documents
Silicone, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
Specifications
- Rør int.Ø:3,00 mm
- Beskrivelse:Size 16, with hose barb of 3,1 mm
- Seq:07
- Størrelse:16