VWR® VisiScope®, CCD Camera Adapters
C-mount Adapter for Visiscope® TL524; focusable C-mount adapter (on upright and inverted microscopes)
Katalog # 630-1749
Leverandør: VWR Collection
Gå til:
Specifikationer
- Beskrivelse:C-mount Adapter for Visiscope® TL524; focusable C-mount adapter (on upright and inverted microscopes)
- Seq:300