VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan objective, 40×/0,60
Catalog # 630-1764
Supplier: VWR Collection
Jump to:
Product Details & Documents
These objectives are designed to be used with inverted microscopes.
- High resolution and contrast
- Ensure field flatness up to F.N. 22, based on infinity-corrected optical system
- Long Working Distance provides a wider working space between the lens surface and the object
Ideal for the observation of culture specimens, clinical examinations and cell testing.
Documents
Avantor Cell Culture Solutions.pdf VWR® for Microscopy
Product Literature
Specifications
- Forstørrelse:40×/0,60
- Beskrivelse:IOS LWD W-Plan objective
- Seq:202