Conductivity standards, Certipur® Supelco®
KCl 0,0001 mol/l, 15 µS/cm, in plastic bottles, 5×100 ml, Plastic bottle, 5
Leverandør: Merck
Gå til:
Product Details & Documents
These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.
- Traceable to PTB and NIST
- Tested against primary reference standards
Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:5
- Beskrivelse:KCl 0,0001 mol/l, 15 µS/cm, in plastic bottles, 5×100 ml
- Cat. No.:1.01811.0105
- M-Clarity quality level:MQ100



