Orders LinkContactUsLinkComponent
Test sieves, US standard
Test sieves, US standard
305 mm, 50 mm, 0,425 mm
Catalog # 44753.NK
Test sieves, US standard
Catalog # 44753.NK
Test sieves, US standard
305 mm, 50 mm, 0,425 mm
Price
3.240,00 kr
Log in for availability
1 * 12 "
Product Details & Documents
The brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments.
  • Superior quality guarantees accuracy and reproducibility
  • Sturdy construction ensures easy stacking
Sieve covers, made of polished brass, have a recessed wire handle.
Specifications
  • Ø:
    305 mm
  • Netstørrelse:
    0,425 mm
  • Højde:
    50 mm
  • Seq:
    54