Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Plastic bottle, 1 L
Katalog # 3727.1000
Ætsende
Leverandør:   Avantor
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Katalog # 3727.1000
Leverandør:  Avantor
CAS nummer:  1310-73-2
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Plastic bottle, 1 L

CAS nummer:  1310-73-2
Price
427,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Caution: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.

Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kogende Pt:119…135 °C (1013 hPa)
  • Smeltende Pt:9…10 °C
  • Tæthed:1,53 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-185-5
  • FN:1824
  • ADR:8,II
Specifikationer

Specifikationstestresultater

  • Assay:
    49.5 - 50.5 %
  • Ammonium Hydroxide Precipitate:
    max. 0,01 %
  • Chloride (Cl):
    max. 0,01 %
  • Heavy Metals (as Ag):
    max. 0,001 %
  • Potassium (K):
    max. 0,01 %
  • Sodium Carbonate (Na₂CO₃):
    max. 0,5 %
  • Sulphate (SO₄):
    max. 0,002 %
  • Trace Impurities (in ppm):
    -
  • Iron (Fe):
    max. 5
  • Nickel (Ni):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5
  • Phosphate (PO₄):
    max. 5

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.