Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Plastic bottle, 2,5 L
Catalog # 3727.2500
Corrosive
Supplier:   Avantor
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Catalog # 3727.2500
Supplier:  Avantor
CAS Number:  1310-73-2
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Plastic bottle, 2,5 L

CAS Number:  1310-73-2
Price
848,00 kr
Log in for availability
1 * 2,5 L
Product Details & Documents

 : IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    49.5 - 50.5 %
  • Ammonium Hydroxide Precipitate:
    max. 0,01 %
  • Chloride (Cl):
    max. 0,01 %
  • Heavy Metals (as Ag):
    max. 0,001 %
  • Potassium (K):
    max. 0,01 %
  • Sodium Carbonate (Na₂CO₃):
    max. 0,5 %
  • Sulphate (SO₄):
    max. 0,002 %
  • Trace Impurities (in ppm):
    -
  • Iron (Fe):
    max. 5
  • Nickel (Ni):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5
  • Phosphate (PO₄):
    max. 5

Specifications

  • Pakning:
    Plastic bottle
  • Pakn:
    2,5 L

Specifications