Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Plastic container, 25 L
Catalog # 3727.9025
Corrosive
Supplier:   Avantor
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Catalog # 3727.9025
Supplier:  Avantor
CAS Number:  1310-73-2
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Natriumhydroxid solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Plastic container, 25 L

CAS Number:  1310-73-2
Price
3.450,00 kr
Log in for availability
1 * 25 L
Product Details & Documents

Caution: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    49.5 - 50.5 %
  • Ammonium Hydroxide Precipitate:
    max. 0,01 %
  • Chloride (Cl):
    max. 0,01 %
  • Heavy Metals (as Ag):
    max. 0,001 %
  • Potassium (K):
    max. 0,01 %
  • Sodium Carbonate (Na₂CO₃):
    max. 0,5 %
  • Sulphate (SO₄):
    max. 0,002 %
  • Trace Impurities (in ppm):
    -
  • Iron (Fe):
    max. 5
  • Nickel (Ni):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5
  • Phosphate (PO₄):
    max. 5

Specifications

  • Pakning:
    Plastic container
  • Pakn:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.