Natriumhydroxid, BAKER ANALYZED® ACS, J.T.Baker®
Plastic bottle for solids, 5 kg
: Avantor
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,08627
- :1390 °C (1013 hPa)
- :323 °C
- :2,13 g/cm³ (20 °C)
- :Stue temperatur
- :215-185-5
- :1823
- :8,II
- :01-2119457892-27
- Assay:min. 97,0 %
- Calcium (Ca):max. 0,005 %
- Chloride (Cl):max. 0,006 %
- Heavy Metals (as Ag):max. 0,001 %
- Magnesium (Mg):max. 0,002 %
- Potassium (K):max. 0,02 %
- Sodium Carbonate (Na2CO3):max. 1,0 %
- Trace Impurities (in ppm):-
- Iron (Fe):max. 3
- Mercury (Hg):max. 0,1
- Nickel (Ni):max. 5
- Nitrogen Compounds (as N):max. 3
- Phosphate (PO4):max. 2
- Sulphate (SO4):max. 30 ppm
- Pakning:Plastic bottle for solids
- Pakn:5 kg



