Natriumhydroxid, BAKER ANALYZED® ACS, J.T.Baker®
25 kg
Katalog # 0348.9025
Leverandør: Avantor
Natriumhydroxid, BAKER ANALYZED® ACS, J.T.Baker®
Katalog # 0348.9025
Leverandør: Avantor
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kogende Pt:1390 °C (1013 hPa)
- Smeltende Pt:323 °C
- Tæthed:2,13 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- NÅ:01-2119457892-27
Documents
Certifikater
Søg efter partinummerspec_BAKR0348.1000.pdf spec_BAKR0348.5000.pdf spec_BAKR0348.9025.pdf
Produktspecifikation
Specifikationer
Specifikationstestresultater
- Assay:min. 97,0 %
- Calcium (Ca):max. 0,005 %
- Chloride (Cl):max. 0,006 %
- Heavy Metals (as Ag):max. 0,001 %
- Magnesium (Mg):max. 0,002 %
- Potassium (K):max. 0,02 %
- Sodium Carbonate (Na2CO3):max. 1,0 %
- Trace Impurities (in ppm):-
- Iron (Fe):max. 3
- Mercury (Hg):max. 0,1
- Nickel (Ni):max. 5
- Nitrogen Compounds (as N):max. 3
- Phosphate (PO4):max. 2
- Sulphate (SO4):max. 30 ppm
Specifikationer
- Pakn:25 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



