Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Electrode cable for plug in head U/plug F, 2×2 mm B, for connecting electrodes with Metrohm plug-in head U to Metrohm instruments (socket F), 1 m
Katalog # METR6.2104.600
Leverandør:   METROHM
Electrode Cables
Katalog # METR6.2104.600
Leverandør:  METROHM
Electrode Cables
Electrode cable for plug in head U/plug F, 2×2 mm B, for connecting electrodes with Metrohm plug-in head U to Metrohm instruments (socket F), 1 m
Price
5.170,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Electrode cable for plug in head U/plug F, 2×2 mm B, for connecting electrodes with Metrohm plug-in head U to Metrohm instruments (socket F)
  • Længde:
    1 m
  • Seq:
    12