Cyclohexan ≥99.8%, SupraSolv® for GC-MS
Glass bottle, 1 L
Leverandør: Merck
Cyclohexan ≥99.8%, SupraSolv® for GC-MS
Katalog # 1.00667.1000
Leverandør: Merck
CAS nummer: 110-82-7
Gå til:
Product Details & Documents
Synonymer: Hexahydrobenzene, Hexamethylene
- CAS nummer:110-82-7
- Formel:C₆H₁₂
- MDL nummer:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Kogende Pt:81 °C (1013 hPa)
- Smeltende Pt:7 °C
- Tæthed:0,7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1145
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-806-2
Documents
Certifikater
Søg efter partinummerMERC1.00667.1000_TDS_EN MERC1.00667.2500_TDS_EN
Product specification
Merck SupraSolv® High Purity Solvents for Gas Chromatography
Product Literature
msds_100667
SDS
Specifikationer
Specifikationstestresultater
- Purity (GC):=99.8%
- Identity(IR):Conforms
- Evaporation Residue:= 3.0 mg/l
- Water:= 0.01%
- Color:=10 Hazen
- GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):= 3ng/ml
- Suitability:Residue Analysis
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



