Orders LinkContactUsLinkComponent
Storage Bags
Storage Bags
Food storage bag, 500×800 mm
Katalog # HKPK100140
Leverandør:   HK PACK KRAUTSCHEID
Storage Bags
Katalog # HKPK100140
Leverandør:  HK PACK KRAUTSCHEID
Storage Bags
Storage Bags
Food storage bag, 500×800 mm
Price
2.270,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents
LDPE transparent bags, ideal for storing and protecting food and small spare parts of all kinds.
  • Food safety is ensured
  • Flexible and tear resistant
  • Freeze-resistant starting 50 μ and heat resistant up to 80 °C
Specifikationer
  • Beskrivelse:
    Food storage bag
  • Dimensioner:
    500×800 mm
  • Seq:
    10