Hydrometers for milk
Hydrometer for milk, 1,015 − 1,045 g/ml, 0,0005 g/ml
Katalog # ALLA2230FC015/20QP
Leverandør: Alla France
Gå til:
Product Details & Documents
Certifications: BS 734
Specifikationer
- Beskrivelse:Hydrometer for milk
- Seq:002
- Inddeling:0,0005 g/ml
- Område:1,015 − 1,045 g/ml