Brefeldin A
Glass bottle for solids, 10 mg
Katalog # CAYM11861-10
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
- CAS nummer:20350-15-6
- Formel:C₁₆H₂₄O₄
- MDL nummer:MFCD00083258
- Molecular Weight:280.4 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 mg



