Brefeldin A
Glass bottle for solids, 5 mg
Katalog # CAYM11861-5
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
- CAS nummer:20350-15-6
- Formel:C₁₆H₂₄O₄
- MDL nummer:MFCD00083258
- Molecular Weight:280.4 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 mg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



