Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ecamsule Related Compounds, Reference Standard

Ecamsule Related Compounds, Reference Standard

Ecamsule Related Compound E, USP reference standard, Vial, 25 mg
Catalog # 1231659.
Supplier:   USP
Ecamsule Related Compounds, Reference Standard
Catalog # 1231659.
Supplier:  USP
CAS Number:  
Ecamsule Related Compounds, Reference Standard

Ecamsule Related Compound E, USP reference standard, Vial, 25 mg

CAS Number:  
Price
9.770,00 kr
Log in for availability
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pakning:
    Vial
  • Pakn:
    25 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Ecamsule Related Compound E
  • Cat. No.:
    1231659.
  • Beskrivelse:
    Ecamsule Related Compound E
  • Synonym(s):
    sodium ((1SR,4SR,E)-3-(4-(hydroxymethyl)benzylidene)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.