Robotic tips for Eppendorf epMotion® automated pipetting system
300 µl, -, -, 10 racks, 96 each
Katalog # 736-0760
Leverandør: Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Katalog # 736-0760
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifikationer
- Pakket:10 racks, 96 each
- Steril:-
- Volumen:300 µl
- Filter:-
- Cat. No.:736-0760
- Miljøcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lav fremstillingspåvirkning