Cedertræsolie
Glass bottle, 25 mL
Katalog # TCIAC0626-25ML
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: Cedar wood oil, Cedar Oil, Cedar Leaf Oil
- CAS nummer:8000-27-9
- MDL nummer:MFCD00132766
- Tæthed:0,98 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationstestresultater
- Appearance:White to Yellow to Orange clear liquid
- Acid Value:max. 20.0
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



